Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5600012

Sigma-Aldrich

Anti-Monomethyl-Histone H3 (Lys18) antibody, Rabbit monoclonal

recombinant, expressed in HEK 293 cells, clone RM167, purified immunoglobulin

Synonym(s):

H3K18me1, Histone H3 (monomethyl K18)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$408.00

$408.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$408.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$408.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

recombinant

expressed in HEK 293 cells

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RM167, monoclonal
recombinant monoclonal

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

ELISA: 0.2-1 μg/mL
western blot: 1-2 μg/mL

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

monomethylation (Lys18)

Gene Information

human ... H3C1(8350)

General description

Histone H3/ histone cluster 1 H3 family member a (HIST1H3A) is a core member of the histone family. HIST1H3A is encoded by the gene mapped to human chromosome 6p22.2. The encoded protein is composed of ~136 amino acids.

Specificity

This antibody reacts to Histone H3 monomethylated at Lysine 18 (K18me1). No cross reactivity with dimethylated Lysine 18 (K18me2) or trimethylated Lysine 18(K18me3), or other methylations in histone H3.

Immunogen

Synthetic peptide corresponding to Monomethyl-Histone H3 (Lys18)

Biochem/physiol Actions

Histones acts as structural scaffolds for DNA packaging. Histone H3 plays an important role in replication and DNA repair. Histone H3 is heavily post-translationally modified than the other members of histone family. Mutations in the gene leads to cerebellar high-grade gliomas. Phosphorylated histone H3 is implicated in the modulation of gene expression and in chromosome condensation/segregation. Monomethyl-histone H3 acts as a potent epigenetic mark for repressed euchromatin in Chlamydomonas.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 50% glycerol, 1% BSA and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Histone H3 phosphorylation: Universal code or lineage specific dialects
Cerutti H, et al.
Epigenetics, 4(2), 71-75 (2009)
Monomethyl Histone H3 Lysine 4 as an Epigenetic Mark for Silenced Euchromatin in Chlamydomonas
Van DK, et al.
Plant Cell, 17(9), 2439-2453 (2005)
Histone H3 K27M mutations in adult cerebellar high-grade gliomas.
Nakata S, et al.
Brain Tumor Pathology, 34(3), 113-119 (2017)
A comprehensive view of the epigenetic landscape part I: DNA methylation, passive and active DNA demethylation pathways and histone variants.
Sadakierska-Chudy A
Neurotoxicity Research, 27(1), 84-97 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service