Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5500136

Sigma-Aldrich

Anti-Lambda Light Chain antibody, Rabbit monoclonal

recombinant, expressed in proprietary host, clone SP147, affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

recombinant

expressed in proprietary host

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

SP147, monoclonal

species reactivity

human (tested)

technique(s)

immunoblotting: 1:100
immunohistochemistry: 1:100

isotype

IgG

shipped in

wet ice

Storage temp.

2-8°C

target post-translational modification

unmodified

General description

Lambda light chains are polypeptide chains located in the cell membrane and cytoplasmic regions of normal B cells and plasma cells. The combination of lambda light chains and heavy chains forms immunoglobulin molecules. There are two classes of light chains found in immunoglobulins, kappa light chains and lambda light chains. Light chain production by lymphoid cells is genetically restricted such that the immunoglobulin molecules produced by an individual cell will only contain a single light chain class. This clonal restriction may be used to indicate the polyclonal or monoclonal nature of B cell and plasma cell populations. The ratio of Kappa to Lambda is about 2:1. The level of kappa or lambda can be greatly elevated in multiple myeloma or other B cell malignancies.

Immunogen

Free lambda light chain from pooled human IgG lambda myeloma sera.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service