Ephrin type-A receptor 2 (EphA2) is a protein-tyrosine kinase in the ephrin receptor subfamily. EphA2 plays a role for angiogenesis and tumor neovascularization. High levels of EphA2 protein expression can be detected in some tumor cells such as breast carcinoma, bladder carcinoma, renal cell carcinoma (RCC), and non-small cell lung carcinoma (NSCLC). Inhibition of EphA2 kinase activity is considered for the treatment of various tumors such as RCC and NSCLC.
Immunogen
Synthetic peptide derived from the internal region of human EphA2 protein.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
0.1 ml rabbit monoclonal antibody purified by protein A/G in PBS/1% BSA buffer pH 7.6 with less than 0.1% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.