Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5300387

Sigma-Aldrich

Monoclonal Anti-RET antibody produced in mouse

clone 8D10C9, ascites fluid

Synonym(s):

RET

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$541.00

$541.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

8D10C9, monoclonal

species reactivity

human

technique(s)

direct ELISA: 1:10,000
western blot: 1:500-1:2,000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RET(5979)

General description

The RET (rearranged during transfection) proto-oncogene is mapped to human chromosome 10q11. It has two isoforms, namely RET9 and RET51 with unique C-terminal tail sequences. RET is made of 20 exons and 21 introns.

Immunogen

Purified recombinant fragment of RET (aa896-1063) expressed in E.coli.
Mouse monoclonal antibody raised against RET

Biochem/physiol Actions

The RET (rearranged during transfection) is involved in signal transduction pathways related to cell proliferation and differentiation. Mutations in RET gene results in medullary thyroid carcinoma and Hirschsprung′s disease. It is involved in normal development and cancer. The isoforms of RET provides distinct protein complexes to promote signals.

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Differential recruitment of E3-ubiquitin ligase complexes regulates RET isoform internalization.
Hyndman BD, et al.
Journal of Cell Science, 130(19), 3282-3296 (2017)
Associations between RET tagSNPs and their haplotypes and susceptibility, clinical severity, and thyroid function in patients with differentiated thyroid cancer.
He C, et al.
PLoS ONE, 12(11), e0187968-e0187968 (2017)
Rui-Xue Huang et al.
International journal of clinical and experimental pathology, 8(5), 5793-5797 (2015-07-21)
The purpose of the study is to investigate the relationship between rs1799939, rs1800858 and rs74799832 polymorphisms of RET with thyroid cancer (TC) susceptibility. Genotypes distribution of control groups were tested by Hardy-Weinberg equilibrium (HWE). Rs1799939, rs1800858 and rs74799832 polymorphisms of
Marjan Zarif Yeganeh et al.
Asian Pacific journal of cancer prevention : APJCP, 16(6), 2107-2117 (2015-04-01)
Thyroid cancer is the most common endocrine neoplasia. The medullary thyroid carcinoma (MTC) is one of the most aggressive forms of thyroid malignancy,accounting for up to 10% of all types of this disease. The mode of inheritance of MTC is
C Tomuschat et al.
Pediatric surgery international, 31(8), 701-710 (2015-07-15)
During the past two decades several genes have been identified that control morphogenesis and differentiation of the enteric neuron system (ENS). These genes, when mutated or deleted, interfere with ENS development. RET gene is the major gene causing Hirschsprung's disease

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service