Matrix metalloproteinase 9 (MMP-9) is also known as gelatinase-B and collagenase type IV-B. It is a proteinase. MMP9 is located on human chromosome 20q13.2.
Immunogen
Purified recombinant fragment of human MMP9 expressed in E.coli. Mouse monoclonal antibody raised against MMP9
Biochem/physiol Actions
Matrix metalloproteinase 9 (MMP-9) regulates inflammatory mediators. It plays a major role in the degradation of extracellular matrix (ECM) components, the formation of chemokine gradients, the generation of reactive oxygen species and the movement of effector cells. It stimulates the progression of autoimmune neo-epitopes. MMP-9 is present at high level in the saliva and tear fluid of patients with Sjoogren′s syndrome.
Physical form
Ascitic fluid containing 0.03% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Matrix metalloproteinase 9 (MMP-9) gene polymorphism and MMP-9 plasma levels in primary Sjogren's syndrome
Hulkkonen J, et al.
Rheumatology (Basel), 43(12), 1476-1479 (2004)
Ureaplasma isolates stimulate pro-inflammatory CC chemokines and matrix metalloproteinase-9 in neonatal and adult monocytes
Glaser, et al.
PLoS ONE, 13(3), e0194514-e0194514 (2018)
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.