Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5300152

Sigma-Aldrich

Monoclonal Anti-KSHV K8? antibody produced in mouse

clone 8C12G10G1, ascites fluid

Synonym(s):

K8

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

8C12G10G1, monoclonal

species reactivity

human

technique(s)

direct ELISA: 1:10,000
immunohistochemistry: 1:200-1:1,000
western blot: 1:500-1:2,000

isotype

IgG2a

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Immunogen

Purified recombinant fragment of KSHV K8 α expressed in E.coli.
Mouse monoclonal antibody raised against KSHV K8α

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu Dai et al.
PloS one, 9(2), e90349-e90349 (2014-03-04)
Kaposi's sarcoma-associated herpesvirus is the causative agent of primary effusion lymphoma (PEL), which arises preferentially in the setting of infection with human immunodeficiency virus (HIV). Even with standard cytotoxic chemotherapy, PEL continues to cause high mortality rates, requiring the development
Guy Journo et al.
Journal of virology, 92(16) (2018-06-15)
Kaposi's sarcoma-associated herpesvirus (KSHV, HHV-8) is a gammaherpesvirus associated with several human malignancies. DNA methylation at CpG dinucleotides is an epigenetic mark dysregulated in many cancer types and in KSHV-infected cells. Several previous studies have analyzed in detail the CpG
Robert Hollingworth et al.
The Journal of general virology, 101(8), 873-883 (2020-06-06)
Gammaherpesviruses establish lifelong latent infection in B lymphocytes and are the causative agent of several B-cell malignancies and lymphoproliferative disorders. While a quiescent latent infection allows these pathogens to evade immune detection, initiation of an alternative lifecycle stage, known as
Robert Hollingworth et al.
Journal of virology, 91(22) (2017-09-01)
Double-strand breaks (DSBs) in DNA are recognized by the Ku70/80 heterodimer and the MRE11-RAD50-NBS1 (MRN) complex and result in activation of the DNA-PK and ATM kinases, which play key roles in regulating the cellular DNA damage response (DDR). DNA tumor
Robert Hollingworth et al.
Viruses, 7(6), 2908-2927 (2015-06-10)
Kaposi's sarcoma-associated herpesvirus (KSHV) is the causative agent of several human malignancies. Human tumour viruses such as KSHV are known to interact with the DNA damage response (DDR), the molecular pathways that recognise and repair lesions in cellular DNA. Here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service