SAB5300011
Monoclonal Anti-THPO antibody produced in mouse
clone 1B11, purified immunoglobulin, buffered aqueous solution
Synonym(s):
MGDF, MKCSF, ML, MPLLG, TPO
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
1B11, monoclonal
form
buffered aqueous solution
mol wt
37.8 kDa
species reactivity
human
technique(s)
direct ELISA: 1:10,000
flow cytometry: 1:200-1:400
western blot: 1:500-1:2,000
isotype
IgG2a
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... THPO(7066)
General description
Immunogen
Mouse monoclonal antibody raised against THPO
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service