SAB5202295
Monoclonal Anti-Citrulline-Atto 595 antibody produced in mouse
clone 6C2.1
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.45
biological source
mouse
conjugate
(Atto 595 Conjugate)
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6C2.1, monoclonal
form
buffered aqueous solution
species reactivity
human
concentration
1.0 mg/mL
technique(s)
immunofluorescence: 1:50
western blot: 1:1000
isotype
IgG1
shipped in
wet ice
storage temp.
−20°C
Immunogen
Synthetic L-Citrulline conjugated to Keyhole Limpet Hemocyanin (KLH).
Physical form
PBS pH 7.4, 50% glyercol, 0.09% Sodium Azide
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service