SAB5202225
Monoclonal Anti-GABA A Receptor-Atto 633 antibody produced in mouse
clone S413-67
About This Item
biological source
mouse
conjugate
Atto 633 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
S413-67, monoclonal
form
buffered aqueous solution
mol wt
60 kDa
species reactivity
rat, human
concentration
1.0 mg/mL
technique(s)
immunofluorescence: 1:100
western blot: 1:1000
isotype
IgG2a
NCBI accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... GABRE(2564)
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service