Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5202073

Sigma-Aldrich

Monoclonal Anti-beta 4 Spectrin-PerCP antibody produced in mouse

clone S393-2, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SPTBN4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$292.00

$292.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$292.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.45

$292.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

conjugate

Peridinin-Chlorophyll-Protein Complex

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

S393-2
monoclonal

description

clone S393-2, purified immunoglobulin, buffered aqueous solution
target molecular weight: 200 kDa

form

buffered aqueous solution

species reactivity

human (Tested), mouse (Predicted), rat (Predicted), African green monkey (Tested)

packaging

pkg of 100 μg

storage condition

avoid repeated freeze/thaw cycles
protect from light

concentration

1.0 mg/mL

technique(s)

immunofluorescence: 1:100
western blot: 1:1000

isotype

IgG1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPTBN4(57731)

Immunogen

Immunogen Description: Fusion protein amino acids 1621-1832 (C-terminal repeats 14 to 15) of human Beta4-spectrin
Immunogen Region: C-terminal
Immunogen Species: Human
Immunogen type: Fusion protein

Application


Western blotting
Dilution 1:1000
Tissue/Cell Type Used: monkey COS-Beta-4-Spectrin-His

Immunofluorescence
Dilution 1:100
Tissue/Cell Type Used: SK-N-BE

Physical form

50% glycerol, 0.1% sodium azide
PBS pH 7.4

Preparation Note

Derived from animal serum

Storage and Stability

Protect from light; avoid repeated freeze-thaw cycles. Store at -20 °C long-term, 2-8 °C short-term.

Other Notes

Additional Product Details:
Does not cross react with other Beta-spectrins
Lot Expiration/Retest Date: Expires 1 year from purchase
Target/Gene Description: Spectrin is the major constituent of the cytoskeletal network underlying the erthrocyte plasma membrane and determines cell shape, arrangement of transmembrane proteins and organization of organelles. It is expressed at very low levels in many tissues, with the strongest expression in the cerebellum, spinal cord, stomach, pituitary gland, liver, pancreas, salivary gland, kidney, bladder and heart . Spectrin beta 4 is a non-erythrocyte member, and is expressed in the brain and pancreatic islets and localisted to the nuclear matrix, cytoplasmic vesicles and PML nuclear bodies. Beta4 spectrins are also essential for membrane stability and the molecular organization of the nodes of Ranvier .
Purification method: Protein G purified
Product type: Primary antibody
Antibody structure: Whole Antibody
Protein ID: NP_066022.2

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service