SAB5201690
Monoclonal Anti-Ataxin 1 - Apc antibody produced in mouse
clone S65-37, purified immunoglobulin
Synonym(s):
Anti-ATX1, Anti-D6S504E, Anti-Spinocerebellar ataxia type 1 protein
Select a Size
$463.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$463.00
Notify Me
Get notified when this item is ready to ship via email.
biological source
mouse
Quality Level
conjugate
Allophycocyanin conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
S65-37, monoclonal
form
buffered aqueous solution
mol wt
antigen predicted mol wt 85 kDa
species reactivity
human, rat, mouse
concentration
1 mg/mL
technique(s)
immunocytochemistry: suitable
western blot: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
mouse ... Atxn1(20238)
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service