Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5201514

Sigma-Aldrich

Monoclonal Anti-Jip2/Ib2 - Pe/Atto 594 antibody produced in mouse

clone S135-37, purified immunoglobulin

Synonym(s):

Anti- C jun amino terminal kinase interacting protein 2, Anti-IB2, Anti-JNK MAP kinase scaffold protein 2, Anti-MAPK8IP2, Anti-PRKM8IPL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

PE/Atto 594 conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S135-37, monoclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt 100 kDa

species reactivity

mouse, rat

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Immunogen

Fusion protein amino acids 226-421 of mouse JIP-2/IB-2. Rat: 94% identity (127/135 amino acids identical). Human: 86% identity (173/200 amino acids identical) >50% identity with JIP-1.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS pH 7.4, 50% glycerol, 0.1% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service