Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5200659

Sigma-Aldrich

Monoclonal Anti-Dicer - Atto 633 antibody produced in mouse

clone S167-7, purified immunoglobulin

Synonym(s):

Anti-Double-strand-specific ribonuclease, Anti-HERNA, Anti-KIAA0928

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

Atto 633 conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S167-7, monoclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt 215 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

mouse ... Dicer1(192119)

Immunogen

Fusion protein amino acids 1638-1899 of mouse Endoribonuclease Dicer

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS pH7.4, 50% glycerol, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service