SAB5200539
Monoclonal Anti-Sur1 - Biotin antibody produced in mouse
clone S289-16, purified immunoglobulin
Synonym(s):
Anti-ABC36, Anti-AM60008PU-N, Anti-Abcc8, Anti-HHF1, Anti-MRP8
Select a Size
$463.00
Notify Me
Get notified when this item is ready to ship via email.
Select a Size
About This Item
$463.00
Notify Me
Get notified when this item is ready to ship via email.
biological source
mouse
Quality Level
conjugate
biotin conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
S289-16, monoclonal
form
buffered aqueous solution
mol wt
antigen predicted mol wt 160 kDa
species reactivity
mouse, rat, hamster
concentration
1 mg/mL
technique(s)
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
rat ... Abcc8(25559)
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service