SAB5200297
Monoclonal Anti-Mmp9 - Atto 565 antibody produced in mouse
clone S51-82, purified immunoglobulin
Synonym(s):
Anti-82kDa matrix metalloproteinase-9, Anti-CLG4B, Anti-GELB, Anti-MANDP2, Anti-MMP-9
About This Item
biological source
mouse
conjugate
Atto 565 conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
S51-82, monoclonal
form
buffered aqueous solution
mol wt
antigen predicted mol wt 92 & 82 kDa
species reactivity
mouse, rat, human
concentration
1 mg/mL
technique(s)
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable
isotype
IgG2a
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Mmp9(81687)
Specificity
Immunogen
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service