Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5200203

Sigma-Aldrich

Monoclonal Anti-Grp78 - Streptavidin antibody produced in mouse

clone 3G12-1G11, purified immunoglobulin

Synonym(s):

Anti-AL022860, Anti-Endoplasmic reticulum lumenal, Anti-HSPA5, Anti-Heat shock 70 kDa protein 5, Anti-MIF2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

streptavidin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G12-1G11, monoclonal

form

buffered aqueous solution

mol wt

antigen predicted mol wt 70 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Specificity

Detects a protein with a mass of ~78kDa corresponding to GRP78

Immunogen

Full-length recombinant rat GRP78

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

PBS pH7.4, 50% glycerol, 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service