Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5200098

Sigma-Aldrich

Anti-AHSA antibody produced in rabbit

whole antiserum

Synonym(s):

Anti-Aha1, Anti-Aha2, Anti-HspC322, Anti-SHSA1, Anti-p38

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$406.00

$406.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
$406.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$406.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

form

aqueous solution

mol wt

antigen predicted mol wt 38 kDa

species reactivity

mouse, rat, human

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Ahsa(217737)

General description

Activator of heat shock protein ATPase 1 (AHSA) is a cochaperone of the heat shock protein 90. The gene ID of the protein is 217737.

Specificity

Detects ~38 kDa.

Immunogen

Mouse Aha1

Biochem/physiol Actions

Activator of heat shock protein ATPase 1 (AHSA) regulates the activity of heat shock protein 90 (HSP90). It does this by bypassing one intermediate in the HSP90 cycle in the absence of ATP.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit antiserum

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fanny Desjardins et al.
Arteriosclerosis, thrombosis, and vascular biology, 32(10), 2484-2492 (2012-08-04)
Vascular endothelial growth factor (VEGF) signaling to endothelial NO synthase (eNOS) plays a central role in angiogenesis. In endothelial cells (ECs), heat-shock protein 90 (Hsp90) is also a regulator of eNOS activity. Our study is designed to determine whether modulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service