Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB5200052

Sigma-Aldrich

Monoclonal Anti-GABRD antibody produced in mouse

clone S151-3, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-GABA-A-receptor-delta, S151-3, Anti-GABAA-RD, Anti-Gabrd, Anti-MGC105467

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S151-3, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 97 kDa

species reactivity

mouse, rat

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Specificity

Apparent mol. wt of 97kD, non-glycosylated version at 68kD. Other minor bands associated with hNIS at160 kDa, and degradation products at ~30 kDa, and ~15 kDa.

Immunogen

Synthetic peptide amino acids 15-34 of rat GABA-A-R-Delta, Accession Number P18506

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingxiao Hong et al.
Frontiers in pharmacology, 11, 612200-612200 (2021-02-09)
Epigenetic modifications such as DNA methylation play important roles in regulating gene expression and may mediate neuroplasticity and lead to drug-induced aberrant behaviors. Although several brain regions and neurobiological mechanisms have been suggested to be involved in these processes, there

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service