Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5200018

Sigma-Aldrich

Monoclonal Anti-PCLO antibody produced in mouse

clone 6H9-B6, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-ACZ, Anti-Aczonin, Anti-PCLO, Anti-PICO, Anti-Piccolo, 6H9-B6, Anti-presynaptic cytomatrix protein antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6H9-B6, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 550 kDa

species reactivity

rat, human

concentration

1 mg/mL

technique(s)

indirect ELISA: suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Pclo(26875)

General description

PCLO (presynaptic cytomatrix protein) gene codes for a protein called piccolo. The encoded protein is a component of the presynaptic active zone.

Specificity

No cross-reactivity to Bassoon, band at ~550 kDa correspondng to the molecular weight of Piccolo.

Immunogen

Full length protein, Accession Number BC001304.1

Biochem/physiol Actions

PCLO (presynaptic cytomatrix protein) gene might be associated with the risk of major depression disorder and bipolar disorder. Mutation in PCLO gene results in pontocerebellar hypoplasia. PCLO serves as a biomarker type 2 diabetes. It controls presynaptic monoamine production. Piccolo plays an important role in synaptic transmission. It mediates the process of docking, vesicle priming and retrieval. Piccolo is involved in neurotransmission and functions at ribbon synapses. Loss of piccolo causes synapse degeneration due to the breakdown of many presynaptic proteins. It regulates ubiquitination, F-actin assembling and proteostasis which helps in maintaining synaptic integrity.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The PCLO gene and depressive disorders: replication in a population-based study
Hek K, et al.
Human Molecular Genetics, 19(4), 731-734 (2009)
Bassoon and Piccolo maintain synapse integrity by regulating protein ubiquitination and degradation
Waites CL, et al.
The Embo Journal, 32(7), 954-969 (2013)
PCLO variants are nominally associated with early onset type 2 diabetes and insulin resistance in Pima Indians
Ma L, et al.
Diabetes (2008)
Gene expression and genetic variation data implicate PCLO in bipolar disorder
Choi KH, et al.
Biological Psychiatry, 69(4), 353-359 (2011)
Mustafa Y Ahmed et al.
Neurology, 84(17), 1745-1750 (2015-04-03)
To identify the genetic cause of pontocerebellar hypoplasia type III (PCH3). We studied the original reported pedigree of PCH3 and performed genetic analysis including genome-wide single nucleotide polymorphism genotyping, linkage analysis, whole-exome sequencing, and Sanger sequencing. Human fetal brain RNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service