Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5200011

Sigma-Aldrich

Monoclonal Anti-YDJ1 antibody produced in mouse

clone 2A7.H6, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-DNAJ1, Anti-DNAJB1, Anti-HDJ1, Anti-HSPF1, Anti-Hsp40, Anti-Hsp40, YDJ1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$541.00

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$541.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$541.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2A7.H6, monoclonal

form

solution

mol wt

antigen predicted mol wt 40 kDa

species reactivity

yeast

concentration

1 mg/mL

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

yeast ... YDJ1(855661)

Specificity

Detects ~40 kDa.

Immunogen

Full length protein Hsp40 (YDJ1)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in 50% glycerol, and 0.09% sodium azide
aqueous glycerol solution

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabian den Brave et al.
Cell reports, 31(9), 107680-107680 (2020-06-04)
The formation of insoluble inclusions in the cytosol and nucleus is associated with impaired protein homeostasis and is a hallmark of several neurodegenerative diseases. Due to the absence of the autophagic machinery, nuclear protein aggregates require a solubilization step preceding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service