Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5100006

Sigma-Aldrich

Imprint® ANTI-POU5F1 (OCT4) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Imprint® ChIP-Validated Anti-POU5F1 (OCT4) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

form

buffered aqueous solution

species reactivity

human

technique(s)

ChIP: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... POU5F1(5460)

Quality

ChIP Validation:

View ChIP Validation Data for lot number: 092M4762v

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as a preservative.

Legal Information

Imprint is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Yang et al.
Bioengineered, 13(1), 1686-1701 (2022-01-11)
Zinc finger protein 267 (ZNF267) is a member of the Kruppel-like transcription factor family, which regulates various biological processes such as cell proliferation and differentiation. However, the biological significance of ZNF267 and its potential role in diffuse large B-cell lymphoma

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service