Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB5100002

Sigma-Aldrich

Imprint® ChIP-Validated Monoclonal Anti-p53 (DO-7) antibody produced in mouse

clone DO-7, purified immunoglobulin, isotype IgG2b

Synonym(s):

Imprint® Monocolonal Anti-p53 antibody produced in mouse, Anti-LFS1, Anti-P53, Anti-TP53, Anti-TRP53, Anti-tumor protein p53

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DO-7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TP53(7157)

General description

The tumor protein p53 (TP53) gene is a tumor suppressor gene that encodes p53 protein. This protein is a nuclear transcription factor. It is made up of three functional domains, such as the NH2-terminal acidic transactivation domain, DNA-binding domain and a COOH-terminal oligomerization domain. p53 protein is located in the nucleus and directly binds to the DNA. The TP53 gene is mapped on the human chromosome at 17p13.1.

Biochem/physiol Actions

p53 protein plays a very important role in suppressing cancer by promoting cell-cycle-arrest or apoptosis as a feedback mechanism to DNA damage. It also plays a role in autophagy, regulating metabolism and oxidative status of the cell. p53 protein is also a part of angiogenesis, senescence, cellular differentiation, and the immune response. Mutations in the TP53 gene are associated with several cancers such as small cell lung cancer, pancreatic cancer, ovarian serous carcinoma, head and neck squamous cell carcinoma and invasive breast carcinoma.

Quality

ChIP Validation:

View ChIP Validation Data for lot number: 032M4762

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as a preservative.

Legal Information

Imprint is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New therapeutic strategies to treat human cancers expressing mutant p53 proteins
Blandino G and Di Agostino S
Journal of Experimental & Clinical Cancer Research, 37(1), 1-13 (2018)
Role of p53 in cell death and human cancers
Ozaki T and Nakagawara A
Cancers, 3(1), 994-1013 (2011)
Recent advances in p53 research and cancer treatment
Suzuki K and Matsubara H
Journal of Biomedicine and Biotechnology, 2011(15), 2527-2532 (2011)
TP53 mutations and SNPs as prognostic and predictive factors in patients with breast cancer
Huszno J and Grzybowska E
Oncology Letters, 16(1), 34-40 (2018)
p53 at a glance
Brady C A and Attardi L D
Journal of Cell Science, 123(15), 2527-2532 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service