Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB5100001

Sigma-Aldrich

Imprint® ChIP-Validated Monoclonal Anti-p53 (DO-1) antibody produced in mouse

clone DO-1, purified immunoglobulin

Synonym(s):

Imprint® Monocolonal Anti-p53 antibody produced in mouse, Anti-LFS1, Anti-P53, Anti-TP53, Anti-TRP53, Anti-tumor protein p53

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

DO-1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TP53(7157)

Looking for similar products? Visit Product Comparison Guide

General description

The optimal amount of antibody per ChIP assay should be empirically determined by the end-user depending upon the amount of chromatin used per reaction. We recommend testing 1-5 μg per ChIP assay.
The tumor suppressor protein p53 responds to diverse cellular stresses to regulate target genes that induce cell cycle arrest, apoptosis, senescence, DNA repair, or changes in metabolism. Wildtype p53 protein is expressed at a low level in normal cells but often mutated and expressed at a high level in a variety of transformed cell lines, where it is believed to contribute to transformation and malignancy. p53 is a DNA-binding protein containing transcription activation, DNA-binding, and oligomerization domains. Mutants of p53 that frequently occur in a number of different human cancers fail to bind the consensus DNA binding site, and hence cause the loss of tumor suppressor activity.

Immunogen

recombinant human wild type p53 1-5.

Application

Suitable for Chip and western blot.

Quality

ChIP Validation

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze at -20°C in working aliquots. Repeated freezing and thawing or storage in “frost-free” freezers is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Other Notes

View ChIP Validation Data for lot number: 032M4761

Legal Information

Imprint is a registered trademark of Sigma-Aldrich Co. LLC
Imprint is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

This product is for R&D use only, not for drug, household, or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jiajia Shen et al.
Cancer research, 78(2), 359-371 (2017-12-01)
As a component of the transcriptional repression complex 1 (PRC1), the ring finger protein RING1 participates in the epigenetic regulation in cancer. However, the contributions of RING1 to cancer etiology or development are unknown. In this study, we report that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service