SAB4800048
Imprint® Monoclonal Anti-Ago antibody produced in mouse
clone 2A8, affinity isolated antibody, buffered aqueous solution
Synonym(s):
Anti-AGO1, Anti-AGO2, Anti-AGO3, Anti-AGO4, Anti-Argonautes
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
2A8, monoclonal
form
buffered aqueous solution
mol wt
antigen ~97 kDa
species reactivity
mouse, human
concentration
~2 μg/μL
technique(s)
RIP: suitable
direct immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
NCBI accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... EIF2C2(27161)
Physical form
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service