Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4800032

Sigma-Aldrich

Imprint® Monoclonal Anti-Ty1 antibody produced in mouse

clone BB2, purified antibody, buffered aqueous solution

Synonym(s):

Anti-Ty1 tag

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

BB2, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~2 μg/μL

technique(s)

ChIP: suitable
western blot: suitable

isotype

IgG1

immunogen sequence

EVHTNQDPLD

shipped in

dry ice

Storage temp.

−20°C

Application

Monoclonal Anti-Ty1 antibody produced in mouse has been used for western blotting.[1]

Physical form

Supplied at 2 mg/mL in PBS.

Legal Information

Imprint is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaomin Shang et al.
Nucleic acids research, 50(6), 3413-3431 (2022-03-16)
Heterochromatin-associated gene silencing controls multiple physiological processes in malaria parasites, however, little is known concerning the regulatory network and cis-acting sequences involved in the organization of heterochromatin and how they modulate heterochromatic gene expression. Based on systematic profiling of genome-wide
Xiaomin Shang et al.
Frontiers in cell and developmental biology, 9, 782293-782293 (2022-01-28)
Plasmodium falciparum undergoes a series of asexual replications in human erythrocytes after infection, which are effective targets for combatting malaria. Here, we report roles of an ApiAP2 transcription factor PfAP2-EXP2 (PF3D7_0611200) in the intraerythrocytic developmental cycle of P. falciparum. PfAP2-EXP2
Anat Florentin et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(24), 13719-13729 (2020-06-03)
The human malaria parasite, Plasmodium falciparum, contains an essential plastid called the apicoplast. Most apicoplast proteins are encoded by the nuclear genome and it is unclear how the plastid proteome is regulated. Here, we study an apicoplast-localized caseinolytic-protease (Clp) system
Martin Zoltner et al.
PLoS neglected tropical diseases, 12(7), e0006679-e0006679 (2018-07-25)
Poly(A)-binding proteins (PABPs) regulate mRNA fate by controlling stability and translation through interactions with both the poly(A) tail and eIF4F complex. Many organisms have several paralogs of PABPs and eIF4F complex components and it is likely that different eIF4F/PABP complex
Yuka Umeki et al.
Cellular and molecular life sciences : CMLS, 80(1), 17-17 (2022-12-24)
Tumors comprise diverse cancer cell populations with specific capabilities for adaptation to the tumor microenvironment, resistance to anticancer treatments, and metastatic dissemination. However, whether these populations are pre-existing in cancer cells or stochastically appear during tumor growth remains unclear. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service