Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

SAB4800023

Sigma-Aldrich

Imprint® Anti-H3S10p antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Histone H3 (phospho S10)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~17 kDa

species reactivity

human

concentration

~1.05 mg/mL

technique(s)

ChIP: suitable
direct immunofluorescence: suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

Physical form

Supplied at 1.05 mg/mL in PBS, 0.05% sodium azide., 0.05% ProClin 300.

Legal Information

Imprint is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service