SAB4800011
Imprint® Monoclonal Anti-TBP, (N-terminal) antibody produced in mouse
purified antibody, buffered aqueous solution
Synonym(s):
Anti-GTF2D, Anti-HDL4, Anti-SCA17, Anti-TATA-box binding protein, Anti-TFIID
About This Item
biological source
mouse
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
form
buffered aqueous solution
mol wt
antigen ~38 kDa
species reactivity
human
concentration
~8 mg/mL
technique(s)
ChIP: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
Gene Information
human ... TBP(6908)
Physical form
Legal Information
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service