Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4701127

Sigma-Aldrich

Monoclonal Anti-CD45R-APC antibody produced in rat

clone RA3-6B2

Synonym(s):

PTPRCR, T200R

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$345.00

$345.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$345.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

$345.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rat

conjugate

Allophycocyanin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

RA3-6B2, monoclonal

form

buffered aqueous solution

concentration

0.5 mg/mL

technique(s)

flow cytometry: 3 μg/test

accession no.

NM_001111316.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Ptprc(19264)

General description

The rat monoclonal antibody RA3-6B2 recognizes an epitope on CD45R, which is expressed at all developmental stages of B cells, including activated B cells, but also on subsets of NK and T cells. T cells detected by this antibody are supposed to be in activated state.

Immunogen

Abelson murine leukemia virus-induced pre-B tumor cells

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service