Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700993

Sigma-Aldrich

Monoclonal Anti-LIME1 antibody produced in mouse

clone LIME-10

Synonym(s):

LITA1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$325.00

$325.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$325.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$325.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LIME-10, monoclonal

form

buffered aqueous solution

concentration

1 mg/mL

technique(s)

western blot: 1 μg/mL

accession no.

NM_001305654.1

shipped in

wet ice

Storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... LIME1(54923)

General description

The antibody LIME-10 reacts with the cytoplasmic domain of LIME, a 30 kDa Lck-interacting transmembrane adaptor expressed by T cells.

Immunogen

COOH-terminal peptide comprising residues 281-296 of the human LIME conjugated to keyhole limpet hemocyanin.

Physical form

Solution in phosphate buffered saline with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service