The mouse monoclonal antibody 10/78 recognizes CD161, an approximately 30 kDa type II transmembrane C-type lectin receptor, expressed on the plasma membrane of NK cells, dendritic cells, activated monocytes and a subset of T cells as a disulphide-linked homodimer. A common epitope on rat CD161a and b isoforms is detected.
Immunogen
Splenic cells purified from the LEW rat
Physical form
Solution in phosphate buffered saline with 15 mM sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.