Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700769

Sigma-Aldrich

Monoclonal Anti-HIV-1 gp24 antibody produced in mouse

clone ND1, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$290.00

$290.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$290.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$290.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ND1, monoclonal

form

buffered aqueous solution

species reactivity

human immunodeficiency virus 1

concentration

1 mg/mL

technique(s)

immunoblotting: suitable
immunocytochemistry: suitable
indirect ELISA: suitable

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

General description

The antibody ND1 reacts with HIV-1 (Human Immunodeficiency Virus Type 1) capsid protein p24 (gp24).
Specificity of antibody ND1 was verified by Western Blotting in reducing conditions with Sanofi Pasteur NEW LAV-BLOT I kit.

Immunogen

HIV-1 p24 C-terminal peptide.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service