Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700644

Sigma-Aldrich

Monoclonal Anti-TCR alpha/beta-Biotin antibody produced in mouse

clone IP26, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TCR α/β

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$248.00

$248.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$248.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$248.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

IP26, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

General description

The mouse monoclonal antibody IP26 recognizes a monomorphic determinant of TCR alpha/beta, the dominant subtype of T cell receptor expressed in human peripheral blood.

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1:300. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service