The antibody SIT-01 reacts with SHP2-interacting transmembrane adaptor protein (SIT) expressed exclusively in lymphoid organs. It weakly crossreacts with mouse SIT.
Immunogen
Bacterially produced recombinant intracellular fragment of human SIT
Application
Suggested working dilution for immunoblotting is 1-5 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.