Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700556

Sigma-Aldrich

Monoclonal Anti-Cd4 azide free antibody produced in rat

clone GK1.5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD4mut, Anti-IMD79, Anti-OKT4D

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$290.00

$290.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$290.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$290.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GK1.5, monoclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... CD4(12504)

General description

Cluster of differentiation 4 (CD4) molecule is encoded by the gene mapped to human chromosome 12p13.31. The encoded protein is mainly localized on the surface of specific leukocytes. CD4 is characterized with a four ecto-domains (D1-D4) of which D1, D2 and D4 contain disulfide bonds.
The rat monoclonal antibody GK1.5 reacts with an extracellular epitope of mouse CD4 transmembrane glycoprotein (55 kDa).

Immunogen

Mouse CTL clone V4 cells

Application

The reagent is designed for Flow Cytometry analysis using 1 μg reagent / 1e6 cells in a suspension.

Biochem/physiol Actions

Cluster of differentiation 4 (CD4) facilitates the activation of immunostimulatory T-cells. It serves as a primary receptor for human immunodeficiency virus (HIV)-1 entry. The encoded protein also plays a key role in altering cell elasticity. Elevated expression of soluble CD4 has been observed in patients of infectious and chronic inflammatory diseases and rheumatoid arthritis (RA). Thus, this protein can be used as a potential diagnostic marker for the development of RA.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isochromosomes 12p and 9p: parental origin and possible mechanisms of formation
Dutly F
European Journal of Human Genetics, 6, 140-144 (1998)
Disease severity in rheumatoid arthritis: relationships of plasma tumor necrosis factor-alpha, soluble interleukin 2-receptor, soluble CD4/CD8 ratio, neopterin, and fibrin D-dimer to traditional severity and functional measures.
Beckham JC
Journal of clinical immunology, 12, 353-361 (1992)
The role of CD4 on mechanical properties of live cell membrane.
Bui VC and Nguyen TH
Journal of Biomedical Materials Research Part A, 104, 239-244 (2016)
Human CD4 Metastability Is a Function of the Allosteric Disulfide Bond in Domain 2.
Owen GR
Biochemistry, 55, 2227-2237 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service