Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700553

Sigma-Aldrich

Monoclonal Anti-Cd3e low endotoxin antibody produced in hamster

clone 145-2C11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CD3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

hamster

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

145-2C11, monoclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Cd3e(12501)

General description

The hamster monoclonal antibody 145-2C11 reacts with mouse CD3 (epsilon subunit). This antibody is commonly used as a phenotypic marker for mouse T cells.

Immunogen

Mouse BM10-37 cytotoxic T lymphocytes

Application

The reagent is designed for Flow Cytometry analysis using 1-2 μg reagent / 1e6 cells in a suspension.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized. Endotoxin level is less than 10 EU/mg of the protein, as determined by the LAL test.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service