The antibody ZAP-03 reacts with ZAP-70, a 70 kDa protein tyrosine kinase expressed in T and NK cells.
Immunogen
Bacterially expressed fusion protein representing C-terminal part (160 amino acids) of human ZAP-70 with histidine tag
Application
The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 2-5 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.