SAB4700495
Monoclonal Anti-CD62P, low endotoxin antibody produced in mouse
clone AK4, purified immunoglobulin, buffered aqueous solution
Synonym(s):
Anti-P-selectin, Anti-SELP
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352203
NACRES:
NA.41
biological source
mouse
conjugate
unconjugated
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
AK4, monoclonal
form
buffered aqueous solution
species reactivity
nonhuman primates, human
concentration
1 mg/mL
technique(s)
flow cytometry: suitable
impurities
≤1 EU/μg endotoxin (LAL test)
isotype
IgG1
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
2-8°C
Gene Information
human ... SELP(6403)
General description
CD62P (P-selectin) is an adhesion glycoprotein that is expressed on platelets and endothelial cells upon their activation. Interaction between CD62P and its mucin-like ligand PSGL-1 (P-selectin glycoprotein ligand-1) expressed on the microvilli of most leukocytes supports leukocyte rolling along postkapillary venules at the earliest time of inflammation. Both CD62P and PSGL-1 are extended glycoproteins that form homodimers. CD62P dimerization is probably mediated through interactions of the transmembrane domains and stabilizes leukocyte tethering and rolling, probably by increasing rebinding within a bond cluster.
Specificity
The antibody AK4 recognizes CD62P (P-selectin), a 140 kD single chain type I transmembrane glycoprotein present in secretory alpha-granules in platelets, in Weibel-Palade bodies in endothelial cells and in megakaryocytes; it is relocated to the plasma membrane upon activation.
Immunogen
Human platelets
Application
The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 1.5 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized. Endotoxin level is less than 0.01 EU/ug of the protein, as determined by the LAL test.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
Storage Class
12 - Non Combustible Liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service