Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700403

Sigma-Aldrich

Monoclonal Anti-CD18, low endotoxin antibody produced in mouse

clone MEM-48, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ITGB2, Anti-Integrin β2 subunit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$218.00

$218.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$218.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$218.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-48, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

impurities

≤1 EU/μg endotoxin (LAL test)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGB2(3689)

Related Categories

General description

The antibody MEM-48 recognizes an epitope involving residues 534-546 in cysteine-rich repeat 3 of the CD18 antigen (integrin beta2 subunit; beta2 integrin). CD18 is a 90-95 kDa type I transmembrane protein expressed on all leukocytes.

Immunogen

Leukocytes of a patient suffering from a LGL-type leukemia

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 2 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized. Endotoxin level is less than 0.01 EU/ug of the protein, as determined by the LAL test.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service