Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700341

Sigma-Aldrich

Monoclonal Anti-CD222 antibody produced in mouse

clone MEM-238, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CIMPR, Anti-IGF2 receptor, Anti-IGF2R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-238, monoclonal

form

buffered aqueous solution

species reactivity

human, nonhuman primates

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

2-8°C

Gene Information

human ... IGF2R(3482)

General description

The antibody MEM-238 recognizes an epitope between domains 2 and 5 of CD222 (IGF2 receptor), a ubiquitously expressed 250 kDa multifunctional type I transmembrane protein. The majority of CD222 is found in the late endosomal/prelysosomal compartment, 5-10% in the plasma membrane and the truncated (220 kDa) form of CD222 is present in human and bovine serum.

Immunogen

Recombinant Vaccinia virus encoding CD222

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution for Flow Cytometry is 1-2 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service