Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700106

Sigma-Aldrich

Monoclonal Anti-CD14-FITC antibody produced in mouse

clone MEM-18, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-LPS-receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

biological source

mouse

conjugate

FITC conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-18, monoclonal

form

buffered aqueous solution

species reactivity

nonhuman primates, human

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD14(929)

General description

CD14 (cluster of differentiation 14) is a myelomonocytic differentiation antigen, encoded by the gene mapped to human chromosome 5q23-31. It is mainly expressed by monocytes, macrophages and activated granulocytes.

Immunogen

A crude mixture of human urinary proteins precipitated by ammonium sulphate from the urine of a patient suffering from proteinuria

Application

The reagent is designed for Flow Cytometry analysis of human blood cells using 20 μL reagent / 100 μL of whole blood or 1e6 cells in a suspension. The content of a vial (2 mL) is sufficient for 100 tests.

Biochem/physiol Actions

CD14 (cluster of differentiation 14) serves as an important receptor of bacterial lipopolysaccharide (LPS). CD14 and LPs complex interacts with toll-like receptor 4 and enhances the expression of inflammatory mediators, including tumor necrosis factor (TNF)- and interleukin (IL)-1, via p38 mitogen-activated protein kinase and nuclear factor-B pathway. Mutation in the gene is associated with the development of myeloid leukemias and generalized chronic periodontitis (GCP).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synergistic contribution of CD14 and HLA loci in the susceptibility to Buerger disease
Chen Z
Human Genetics, 122, 367-372 (2007)
Cytogenetic and molecular delineation of the smallest commonly deleted region of chromosome 5 in malignant myeloid diseases
Le Beau MM
Proceedings of the National Academy of Sciences of the USA, 90, 5484-5488 (1993)
Dilshi Ishaan et al.
Genetic testing and molecular biomarkers, 21(9), 560-564 (2017-08-23)
The genetic variation in the CD14 rs2569190 promoter region contributes to the susceptibility of developing periodontitis. Since, CD14 gene polymorphism studies among various ethnic populations have produced conflicting results, the present study was designed to unearth the association between the
The CD14 monocyte differentiation antigen maps to a region encoding growth factors and receptors.
Goyert SM
Science, 239, 497-500 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service