Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4700024

Sigma-Aldrich

Monoclonal Anti-CD147-Biotin antibody produced in mouse

clone MEM-M6/1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BSG, Anti-BSG-Biotin, Anti-Basigin, Anti-Neurothelin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

biotin conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-M6/1, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

1 mg/mL

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... BSG(682)

General description

The antibody MEM-M6/1 recognizes an epitope in the N-terminal Ig domain (D1) of CD147 (Neurothelin), a 50-60 kDa type I transmembrane glycoprotein primarily expressed on all leukocytes, red blood cells, platelets and endothelial cells; it is not expressed by resting lymphocytes.
Cluster of differentiation 147 (CD147) is a multifunctional trans-membrane glycoprotein and is a member of the immunoglobulin superfamily. It occurs in transmembrane and soluble forms. CD147 is also known as EMMPRIN or basigin (BSG). It is a 57 kDa glycoprotein with two extracellular Ig domains. CD147 is located on human chromosome 19p13.

Immunogen

Protein A-CR purified soluble recombinant form of CD147, CD147Rg, which consists of the cDNA coding for the hinge region, CH2-and CH3 domain of human IgG1 (CD147Rg is secreted by transfectants as a dimer)

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1:250. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Biochem/physiol Actions

Cluster of differentiation 147 (CD147) is considered as an extracellular matrix metalloproteinase inducer. It participates in several physiological and pathological activities, mainly in tumor invasion and metastasis. CD147 helps in tumor cell-macrophage interactions and stimulates MMPs (matrix metalloproteinases) and vascular endothelial growth factor (VEGF).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cytoplasmic fragment of CD147 generated by regulated intramembrane proteolysis contributes to HCC by promoting autophagy.
Wu B, et al.
Cell Death & Disease, 8(7), e2925-e2925 (2017)
CD147 overexpression may serve as a promising diagnostic and prognostic marker for gastric cancer: evidence from original research and literature.
Hu C, et al.
Oncotarget, 8(19), 30888-30888 (2017)
CD147-induced cell proliferation is associated with Smad4 signal inhibition.
Qin H, et al.
Experimental Cell Research, 358(2), 279-289 (2017)
BGMUT: NCBI dbRBC database of allelic variations of genes encoding antigens of blood group systems.
Patnaik SK, et al.
Nucleic Acids Research, 40(D1), D1023-D1029 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service