Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4600220

Sigma-Aldrich

Anti-Rat IgG (H+L), highly crossed-adsorbed, CF350 antibody produced in donkey

~2 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

250 μL
$221.00

$221.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
250 μL
$221.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

$221.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

donkey

Quality Level

conjugate

CF 350 conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat

concentration

~2 mg/mL

technique(s)

flow cytometry: 1-10 μg/mL
immunocytochemistry: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
indirect immunofluorescence: 1-10 μg/mL
western blot: suitable

fluorescence

λex 347 nm; λem 448 nm

storage temp.

−20°C

target post-translational modification

unmodified

General description

This product is prepared by labeling highly cross-adsorbed donkey anti-rabbit IgG (H+L) with CF350. CF350 is a blue fluorescent dye spectrally similar to the traditional blue fluorescent dye, AMCA. However, compared to AMCA, CF350 is more water-soluble, more photostable and at least 50% more fluorescent on proteins. CF350 is at least as bright as Alexa fluor 350 and thus can be a direct replacement for the latter for all applications. To minimize cross-reactivity, the antibody has been adsorbed against Bovine, Chicken, Goat, Guinea Pig, Syrian Hamster, Horse, Human, Mouse, Rabbit, and Sheep serum.

Specificity

Binds all rat IgGs (minimal cross-reaction with bovine, chicken, goat, guinea pig, hamster, horse, human, mouse, rabbit and sheep serum proteins)

Immunogen

rat IgG(H+L)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied in phosphate buffered saline with 0.05% sodium azide, 50% glycerol and 0.2% BSA.

Preparation Note

Protect from light. The antibody solution should be gently mixed before use.

Legal Information

This product is distributed by Sigma-Aldrich Co. under the authorization of Biotium, Inc. This product is covered by one or more US patents and corresponding patent claims outside the US patents or pending applications owned or licensed by Biotium, Inc. including without limitation: 12/334,387; 12/607,915; 12/699,778; 12/850,578; 61/454,484. In consideration of the purchase price paid by the buyer, the buyer is hereby granted a limited, non-exclusive, non-transferable license to use only the purchased amount of the product solely for the buyer′s own internal research in a manner consistent with the accompanying product literature. Except as expressly granted herein, the sale of this product does not grant to or convey upon the buyer any license, expressly, by implication or estoppel, under any patent right or other intellectual property right of Biotium, Inc. Buyer shall not resell or transfer this product to any third party, or use the product for any commercial purposes, including without limitation, any diagnostic, therapeutic or prophylactic uses. This product is for research use only. Any other uses, including diagnostic uses, require a separate license from Biotium, Inc. For information on purchasing a license to use this product for purposes other than research, contact Biotium, Inc., 3159 Corporate Place, Hayward, CA 94545, Tel: (510) 265-1027. Fax: (510) 265-1352. Email: [email protected].
CF is a trademark of Biotium, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu-Ming Li et al.
Developmental dynamics : an official publication of the American Association of Anatomists, 253(5), 467-489 (2023-10-18)
The environmental oxygen tension has been reported to impact the blastocyst quality and cell numbers in the inner cell mass (ICM) during human and murine embryogenesis. While the molecular mechanisms leading to increased ICM cell numbers and pluripotency gene expression

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service