Skip to Content
MilliporeSigma
All Photos(6)

Key Documents

SAB4504739

Sigma-Aldrich

Anti-phospho-Zyxin (pSer142) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 61 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer142)

Gene Information

human ... ZYX(7791)

General description

The ZYX (zyxin) gene is mapped to human chromosome 7q34. It encodes a zinc-binding phosphoprotein, belonging to the focal adhesion complex. It is also localized to actin cytoskeleton.

Immunogen

The antiserum was produced against synthesized peptide derived from human Zyxin around the phosphorylation site of Ser142.

Immunogen Range: 108-157

Biochem/physiol Actions

ZYX (zyxin) controls actin filament assembly, and cell motility. It also participates in the regulation of cell division (mitosis), and cell differentiation processes. ZYX is associated with tumor cell proliferation and migration. It is also known to induce the pathogenesis of colon cancer, and hepatocellular carcinoma. Mechanoresponses with respect to integrin and cadherin-based cell adhesion, shows the involvement of zyxin.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VASP, zyxin and TES are tension-dependent members of Focal Adherens Junctions independent of the alpha-catenin-vinculin module
Oldenburg J, et al.
Scientific Reports, 5(29), 17225-17225 (2015)
Zyxin promotes colon cancer tumorigenesis in a mitotic phosphorylation-dependent manner and through CDK8-mediated YAP activation
Zhou J, et al.
Proceedings of the National Academy of Sciences of the USA, 115(29), E6760-E6769 (2018)
Novel identification of zyxin upregulations in the motile phenotype of hepatocellular carcinoma
Sy SMH, et al.
Modern Pathology, 19(8), 1108-1108 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service