Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4504199

Sigma-Aldrich

Anti-phospho-JIP1 (pThr103) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 77 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

Gene Information

human ... MAPK8IP1(9479)

General description

Mitogen-activated protein kinase 8 interacting protein 1/JNK interacting protein 1 (JIP-1) is a pivotal scaffolding protein. It is also termed as islet brain 1 (IB1). It is present in the brain and islets. JIP1 gene is located on human chromosome 11p11.

Immunogen

The antiserum was produced against synthesized peptide derived from human JIP1 around the phosphorylation site of Thr103.

Immunogen Range: 69-118

Biochem/physiol Actions

Mitogen-activated protein kinase 8 interacting protein 1/JNK interacting protein 1 (JIP-1) controls the c-Jun N-terminal kinase (JNK) activity involved in cell differentiation, division and death. This gene may help to mediate mitogenic messages from the plasma membrane to the cell interior. JIP-1 also controls the JNK pathway in neuronal and beta-cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of the JNK-interacting protein 1/islet brain 1 in cell degeneration in Alzheimer disease and diabetes
Beeler N, et al.
Brain Research Bulletin, 80(4-5), 274-278 (2009)
The JNK interacting protein JIP-1 and insulin like growth factor II genes are co-expressed in human embryonic tumours
Engstrom W, et al.
Anticancer Research (2005)
Proximal 11p deletion syndrome (P11pDS): additional evaluation of the clinical and molecular aspects
Wuyts W, et al.
European Journal of Human Genetics, 12(5), 400-406 (2004)
IB1/JIP-1 controls JNK activation and increased during prostatic LNCaP cells neuroendocrine differentiation
Tawadros T, et al.
Cellular Signalling, 17(8), 929-939 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service