SAB4504055 has been used in Immunohistochemistry and has passed our specification for
1:50~1:100. We have also found reactivity in mouse. However, we have not studied this with mouse tumor samples. In searching both PubMed and Google Scholar for you, there are no references to this application with this product.
SAB4504055
Anti-phospho-Pim-1 (pTyr309) antibody produced in rabbit
affinity isolated antibody
Select a Size
$522.00
Select a Size
About This Item
$522.00
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 45 kDa
species reactivity
rat, human, mouse
concentration
~1 mg/mL
technique(s)
ELISA: 1:40000
western blot: 1:500-1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
phosphorylation (pTyr309)
Gene Information
human ... PIM1(5292)
Immunogen
Immunogen Range: 281-330
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Can the antibody phospho PIM1 Tyr 309 SAB4504055 be used for Immunohistochemistry on mouse tumor samples?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service