Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4503819

Sigma-Aldrich

Anti-phospho-Filamin A (pSer2152) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ABP-280, Anti-ABPX, Anti-CSBS, Anti-CVD1, Anti-FGS2, Anti-FLN, Anti-FLN-A, Anti-FMD, Anti-NHBP, Anti-OPD, Anti-OPD1, Anti-OPD2, Anti-XLVD, Anti-XMVD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 280 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FLNA(2316)

General description

Filamin A (FLNA) is an X-linked, ubiquitous actin-binding protein. It is a 280kDa protein consisting of two N-terminal calponin homology domains and a long C-terminal rod-like domain. FLNA is mapped to human chromosome Xq28.

Immunogen

The antiserum was produced against synthesized peptide derived from human Filamin A around the phosphorylation site of Ser2152.

Immunogen Range: 2121-2170

Biochem/physiol Actions

Filamin A (FLNA) has the ability to stop or induce the growth and development of cancer. It regulates the gene transcription of RNA polymerase III in transformed cell lines. In males, mutation in filamin A (FLNA) results in Ehlers-Danlos syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abnormal development of the human cerebral cortex: genetics, functional consequences and treatment options
Guerrini R, et al.
Trends in Neurosciences, 31(3), 154-162 (2008)
Cytoskeletal Filamin A Differentially Modulates RNA Polymerase III Gene Transcription in Transformed Cell Lines
Wang J, et al.
The Journal of Biological Chemistry, 291(48), 25239-25246 (2016)
Ehlers-Danlos syndrome with lethal cardiac valvular dystrophy in males carrying a novel splice mutation in FLNA
Ritelli M, et al.
American Journal of Medical Genetics. Part A, 173(1), 169-176 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service