SAB4503682
Anti-ARHGEF10 antibody produced in rabbit
affinity isolated antibody
Synonym(s):
ARHGEF10, Rho guanine nucleotide exchange factor 10
Select a Size
$392.00
Select a Size
About This Item
$392.00
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 151 kDa
species reactivity
human
concentration
~1 mg/mL
technique(s)
ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... ARHGEF10(9639)
Related Categories
General description
The gene ARHGEF10 (Rho guanine nucleotide exchange factor 10) is mapped to human chromosome 8p23. It belongs to the Rho-specific guanine nucleotide exchange factor family. The protein contains tandem Dbl homology (DH) domain, pleckstrin homology (PH) domain and putative transmembrane domains.
Immunogen
Immunogen Range: 339-388
Biochem/physiol Actions
Features and Benefits
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service