Anti-DGKI Antibody detects endogenous levels of total DGKI protein.
DGKI (Diacylglycerol kinase iota) is a small, integral membrane protein belonging to the type IV diacylglycerol kinase subfamily. It is mapped on chromosome 7q32.3-q33. The members of the DGK family consist of a catalytic domain and at least two cysteine-rich regions. In type IV family members, it has four ankyrin repeats and a carboxy terminal PDZ binding domain.
Immunogen
The antiserum was produced against synthesized peptide derived from human DGKI.
Immunogen Range: 991-1040
Application
Anti-DGKI, C-Terminal antibody produced in rabbit is suitable for indirect immunofluorescence and western blot.
Biochem/physiol Actions
DGKI (Diacylglycerol kinase iota) is mainly involved in the maintenance of intracellular concentration of diacylglycerol by phosphorylating other lipids in addition to DAG. DGKI has been predicted to be a potential gene for tumor suppression in colorectal cancers and mutation in DGKI has been linked to the melanoma.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Biochimica et biophysica acta, 1790(6), 416-424 (2009-04-15)
The mammalian diacylglycerol kinases (DGK) are a group of enzymes having important roles in regulating many biological processes. Both the product and the substrate of these enzymes, i.e. diacylglycerol and phosphatidic acid, are important lipid signalling molecules. Each DGK isoform
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.