Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4503278

Sigma-Aldrich

Anti-CDCA3, C-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

C8, Cell division cycle-associated protein 3, Gene-rich cluster protein C8, TOME-1, Trigger of mitotic entry protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 28 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDCA3(83461)

General description

Anti-CDCA3 Antibody detects endogenous levels of total CDCA3 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CDCA3.

Immunogen Range: 219-268

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R C Zou et al.
Neoplasma, 67(6), 1223-1232 (2020-07-24)
Downregulation of cell division cycle-associated 3 (CDCA3) markedly inhibited cell growth and induced apoptosis in tumors. However, the effect of CDCA3 in pancreatic cancer (PAC) was rarely investigated. Therefore, this study attempted to clarify the role of CDCA3 in PAC.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service