Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4503206

Sigma-Aldrich

Anti-T3JAM antibody produced in rabbit

affinity isolated antibody

Synonym(s):

T3JAM, TRAF3 interacting protein 3, TRAF3-interacting JNK-activating modulator, TRAF3-interacting Jun N-terminal kinase (JNK)-activating modulator, TRAF3-interacting protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 63 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Anti-T3JAM Antibody detects endogenous levels of total T3JAM protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human T3JAM.

Immunogen Range: 251-300

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chin-Yi Cheng et al.
Chinese medicine, 16(1), 82-82 (2021-08-23)
Post-ischemic inflammation is a crucial component in stroke pathology in the early phase of cerebral ischemia-reperfusion (I/R) injury. Inflammation caused by microglia, astrocytes, and necrotic cells, produces pro-inflammatory mediators and exacerbates cerebral I/R injury. This study evaluated the effects of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service